missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dengue Virus Dengue Envelope-2 Protein
A cDNA sequence encoding the Dengue Envelope-2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11626-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Dengue Envelope-2 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
Dengue Envelope-2 protein solution in 1xD-PBS, pH 7.4, 0.1% Thimerosal, 5mM EDTA, 1microg/ml of Leupeptin, Aprotinin and Pepstatin A. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Dengue Virus | |
Untagged | |
Protein is >95% pure as determined by 12.5% SDS-PAGE. |