missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Dog Annexin A13 Protein
A DNA sequence encoding the Canis lupus familiaris (Dog) Annexin A13, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8980-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001003255.1 | |
Annexin A13 Protein | |
Research Use Only | |
ANXA13 | |
Recombinant Protein | |
Yeast | |
GNRHAKAKSH HGFDVDHDAK KLNKACKGMG TDEAAIIEIL SSRTSDERQQ IKQKYKATYG KDLEEVFKSD LSGNFEKTAL ALLDRPSEYD ARQLQKAMKG LGTDEAVLIE ILCTRTNKEI MAIKEAYQRL FDRSLESDVK ADTSGNLKAI LVSLLQANRD EGDDVDKDLA GQDAKDLYDA GDGRWGTDEL AFNEVLAKRS HKQLRATFQA YQILIDKDIE EAIEAETSGD LQKAYLTLVR CARDQEGYFA DRLYKSMKGT GTDEETLIHI IVTRAEVDLQ GIKAKFQEKY QKSLSDMVRS DTSGDFQKLL VALLH | |
Greater than 90% as determined by SDS-PAGE. |
403935 | |
50 μg | |
Not determined. | |
Full Length | |
Dog | |
His | |
Tris-based buffer, 50% glycerol |