missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli Cell division protein ZipA Protein
A DNA sequence encoding the Escherichia coli (strain K12) Cell division protein ZipA, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6957-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_416907.1 | |
Cell division protein ZipA Protein | |
Research Use Only | |
zipA | |
Recombinant Protein | |
E. coli | |
MMQDLRLILI IVGAIAIIAL LVHGFWTSRK ERSSMFRDRP LKRMKSKRDD DSYDEDVEDD EGVGEVRVHR VNHAPANAQE HEAARPSPQH QYQPPYASAQ PRQPVQQPPE AQVPPQHAPH PAQPVQQPAY QPQPEQPLQQ PVSPQVAPAP QPVHSAPQPA QQAFQPAEPV AAPQPEPVAE PAPVMDKPKR KEAVIIMNVA AHHGSELNGE LLLNSIQQAG FIFGDMNIYH RHLSPDGSGP ALFSLANMVK PGTFDPEMKD FTTPGVTIFM QVPSYGDELQ NFKLMLQSAQ HIADEVGGVV LDDQRRMMTP QKLREYQDII REVKDANA | |
Greater than 90% as determined by SDS-PAGE. |
946869 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |