Learn More
enQuireBio™ Recombinant E. coli Glutamate-pyruvate aminotransferase AlaC Protein
A DNA sequence encoding the Escherichia coli (strain K12) Glutamate-pyruvate aminotransferase AlaC, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6950-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_416880.1 | |
Glutamate-pyruvate aminotransferase AlaC Protein | |
Research Use Only | |
alaC | |
Recombinant Protein | |
E. coli | |
MADTRPERRF TRIDRLPPYV FNITAELKMA ARRRGEDIID FSMGNPDGAT PPHIVEKLCT VAQRPDTHGY STSRGIPRLR RAISRWYQDR YDVEIDPESE AIVTIGSKEG LAHLMLATLD HGDTVLVPNP SYPIHIYGAV IAGAQVRSVP LVEGVDFFNE LERAIRESYP KPKMMILGFP SNPTAQCVEL EFFEKVVALA KRYDVLVVHD LAYADIVYDG WKAPSIMQVP GARDVAVEFF TLSKSYNMAG WRIGFMVGNK TLVSALARIK SYHDYGTFTP LQVAAIAALE GDQQCVRDIA EQYKRRRDVL VKGLHEAGWM VEMPKASMYV WAKIPEPYAA MGSLEFAKKL LNEAKVCVSP GIGFGDYGDT HVRFALIENR DRIRQAIRGI KAMFRADGLL PASSKHIHEN AE | |
Greater than 90% as determined by SDS-PAGE. |
946850 | |
500 μg | |
Not determined. | |
Full Length | |
E. coli | |
His-SUMO | |
Tris-based buffer, 50% glycerol |