missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. Coli LACTB Protein
A cDNA sequence encoding the LACTB was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12522-10mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
948669 | |
10 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MAPQQINDIV HRTITPLIEQ QKIPGMAVAV IYQGKPYYFT WGYADIAKKQ PVTQQTLFEL GSVSKTFTGV LGGDAIARGE IKLSDPTTKY WPELTAKQWN GITLLHLATY TAGGLPLQVP DEVKSSSDLL RFYQNWQPAW APGTQRLYAN SSIGLFGALA VKPSGLSFEQ AMQTRVFQPL KLNHTWINVP PAEEKNYAWG YREGKAVHVS PGALDAEAYG VKSTIEDMAR WVQSNLKPLD INEKTLQQGI QLAQSRYWQT GDMYQGLGWE MLDWPVNPDS IINGSDNKIA LAARPVKAIT PPTPAVRASW VHKTGATGGF GSYVAFIPEK ELGIVMLANK NYPNPARVDA AWQILNALQ | |
Greater than 95.0% as determined by SDS-PAGE. |
LACTB Protein | |
Research Use Only | |
ampC | |
E. coli | |
Untagged | |
The LACTB enzyme (1 mg/ml) is supplied in 20mM Tris-HCl buffer (pH 8.0) and 10% glycerol. |