missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant E. coli UDP-glucose 6-dehydrogenase Protein
A DNA sequence encoding the Escherichia coli (strain K12) UDP-glucose 6-dehydrogenase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8889-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_416532.1 | |
UDP-glucose 6-dehydrogenase Protein | |
Research Use Only | |
ugd | |
Recombinant Protein | |
E. coli | |
KITISGTGYV GLSNGLLIAQ NHEVVALDIL PSRVAMLNDR ISPIVDKEIQ QFLQSDKIHF NATLDKNEAY RDADYVIIAT PTDYDPKTNY FNTSSVESVI KDVVEINPYA VMVIKSTVPV GFTAAMHKKY RTENIIFSPE FLREGKALYD NLHPSRIVIG ERSERAERFA ALLQEGAIKQ NIPMLFTDST EAEAIKLFAN TYLAMRVAYF NELDSYAESL GLNSRQIIEG VCLDPRIGNH YNNPSFGYGG YCLPKDTKQL LANYQSVPNN LISAIVDANR TRKDFIADAI LSRKPQVVGI YRLIMKSGSD NFRASSIQGI MKRIKAKGVE VIIYEPVMKE DSFFNSRLER DLATFKQQAD VIISNRMAEE LKDVADKVYT RDLFGSD | |
Greater than 90% as determined by SDS-PAGE. |
946571 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
E. coli | |
His | |
Tris-based buffer, 50% glycerol |