missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV Core Genotype-3b Protein
A cDNA sequence encoding the HCV Core Genotype-3b was constructed and used to recombinantly synthesize the protein.
205.00€ - 1600.00€
Specifica
Nome | Hepatitis C HCV Core Genotype-3b Protein |
---|---|
Purezza | HCV Core Genotype-3b protein is >95% pure as determined by 10% PAGE (coomassie staining). |
Status giuridico | Research Use Only |
Concentrazione di endotossine | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
Product Type | Recombinant Protein |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
15989528
|
enQuireBio™
QP12152-100UG |
100 μg |
205.00€
100µg |
Eseguire il login per visualizzare lo stock disponibile |
15909538
|
enQuireBio™
QP12152-500UG |
500 μg |
800.00€
500µg |
Eseguire il login per visualizzare lo stock disponibile |
15999528
|
enQuireBio™
QP12152-1MG |
1 mg |
1600.00€
1mg |
Eseguire il login per visualizzare lo stock disponibile |
Specifica
Hepatitis C HCV Core Genotype-3b Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-Hcl pH 8.0, 0.2% Triton-X, 1.5M urea and 50% glycerol. |
HCV Core Genotype-3b protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
GST |