missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C HCV NS5B Protein
A cDNA sequence encoding the HCV NS5B was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12200-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hepatitis C HCV NS5B Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis C | |
His |
Protein is >95% pure as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
1.5M urea, 25mM Tris-HCl pH 8.0, 0.2% Triton-X and 50% Glycerol. |