missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis C virus genotype 1a (isolate 1) polyProtein
A DNA sequence encoding the Hepatitis C virus genotype 1a (isolate 1) (HCV) Hepatitis C virus genotype 1a (isolate 1) polyprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6996-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hepatitis C virus genotype 1a (isolate 1) polyProtein | |
Research Use Only | |
Partial (see sequence information for more details). | |
Hepatitis | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
500 μg | |
Not determined. | |
Recombinant Protein | |
Mammal | |
YQVRNSTGLY HVTNDCPNSS IVYEAADAIL HTPGCVPCVR EGNASRCWVA MTPTVATRDG KLPATQLRRH IDLLVGSATL CSALYVGDLC GSVFLVGQLF TFSPRRHWTT QGCNCSIYPG HITGHRMAWD MMMN | |
Greater than 90% as determined by SDS-PAGE. |