missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Hepatitis E HEV ORF2 Protein
A cDNA sequence encoding the HEV ORF2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12222-BG-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
Hepatitis E HEV ORF2 Protein | |
500 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Hepatitis E | |
Beta-galactosidase |
HEV ORF2 protein is >95% pure as determined by 10% PAGE (coomassie staining). | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
25mM Tris-HCl, 1mM EDTA, 0.5M urea and 50% glycerol. |