missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACBD4 Protein
A DNA sequence encoding the Homo sapiens (Human) ACBD4, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7825-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001129177.1 | |
ACBD4 Protein | |
Research Use Only | |
ACBD4 | |
Recombinant Protein | |
E. coli | |
MGTEKESPEP DCQKQFQAAV SVIQNLPKNG SYRPSYEEML RFYSYYKQAT MGPCLVPRPG FWDPIGRYKW DAWNSLGKMS REEAMSAYIT EMKLVAQKVI DTVPLGEVAE DMFGYFEPLY QVIPDMPRPP ETFLRRVTGW KEQVVNGDVG AVSEPPCLPK EPAPPSPESH SPRDLDSEVF CDSLEQLEPE LVWTEQRAAS GGKRDPRNSP VPPTKKEGLR GSPPGPQELD VWLLGTVRAL QESMQEVQAR VQSLESMPRP PEQRPQPRPS ARPWPLGLPG PALLFFLLWP FVVQWLFRMF RTQKR | |
Greater than 90% as determined by SDS-PAGE. |
79777 | |
500 μg | |
Not determined. | |
Full Length of Isoform 2 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |