missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACBD6 Protein
A cDNA sequence encoding the ACBD6 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10938-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
ACBD6 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMASSFLP AGAITGDSGG ELSSGDDSGE VEFPHSPEIE ETSCLAELFE KAAAHLQGLI QVASREQLLY LYARYKQVKV GNCNTPKPSF FDFEGKQKWE AWKALGDSSP SQAMQEYIAV VKKLDPGWNP QIPEKKGKEA NTGFGGPVIS SLYHEETIRE EDKNIFDYCR ENNIDHITKA IKSKNVDVNV KDEEGRALLH WACDRGHKEL VTVLLQHRAD INCQDNEGQT ALHYASACEF LDIVELLLQS GADPTLRDQD GCLPEEVTGC KTVSLVLQRH TTGKA | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
ACBD6 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |