missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ACP5 Protein
A cDNA sequence encoding the ACP5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10502-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
54 | |
10 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Specific activity is >10,000 units/mg, and is defined as the amount of enzyme that hydrolyze 1.0 nmoles of p-nitrophenyl phosphate (pNPP) per minute at pH 5.0 at 37C. | |
Human | |
Untagged | |
ACP5 protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
ACP5 Protein | |
Research Use Only | |
ACP5 | |
Recombinant Protein | |
Baculovirus | |
ATPALRFVAV GDWGGVPNAP FHTAREMANA KEIARTVQIL GADFILSLGD NFYFTGVQDI NDKRFQETFE DVFSDRSLRK VPWYVLAGNH DHLGNVSAQI AYSKISKRWN FPSPFYRLHF KIPQTNVSVA IFMLDTVTLC GNSDDFLSQQ PERPRDVKLA RTQLSWLKKQ LAAAREDYVL VAGHYPVWSI AEHGPTHCLV KQLRPLLATY GVTAYLCGHD HNLQYLQDEN GVGYVLSGAG NFMDPSKRHQ RKVPNGYLRF HYGTEDSLGG FAYVEISSKE MTVTYIEASG KSLFKTRLPR RARP | |
Greater than 90.0% as determined by SDS-PAGE. |