missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human ADSL Protein
A cDNA sequence encoding the ADSL was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10973-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
ADSL Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMAAG GDHGSPDSYR SPLASRYASP EMCFVFSDRY KFRTWRQLWL WLAEAEQTLG LPITDEQIQE MKSNLENIDF KMAAEEEKRL RHDVMAHVHT FGHCCPKAAG IIHLGATSCY VGDNTDLIIL RNALDLLLPK LARVISRLAD FAKERASLPT LGFTHFQPAQ LTTVGKRCCL WIQDLCMDLQ NLKRVRDDLR FRGVKGTTGT QASFLQLFEG DDHKVEQLDK MVTEKAGFKR AFIITGQTYT RKVDIEVLSV LASLGASVHK ICTDIRLLAN LKEMEEPFEK QQIGSSAMPY KRNPMRSERC CSLARHLMTL VMDPLQTASV QWFERTLDDS ANRRICLAEA FLTADTILNT LQNISEGLVV YPKVIERRIR QELPFMATEN IIMAMVKAGG SRQDCHEKIR VLSQQAASVV KQEGGDNDLI ERIQVDAYFS PIHSQLDHLL DPSSFTGRAS QQVQRFLEEE VYPLLKPYES VMKVKAELCL | |
Greater than 95.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The ADSL solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 40% glycerol and 0.1M NaCl. |