missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AKR7A2 Protein
A cDNA sequence encoding the AKR7A2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10514-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
AKR7A2 Protein | |
Research Use Only | |
Specific activity: approximately 0.25-0.3 units/mg. Enzymatic activity was confirmed by measuring the amount of enzyme catalyzing the oxidation of 1 micromole NADPH per minute at 25C. Specific activity was expressed as units/mg protein. | |
Human | |
His | |
The AKR7A2 solution contains 20mM Tris-HCl pH 8.0, 1mM DTT and 20% glycerol. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSELEM LSAASRVVSR AAVHCALRSP PPEARALAMS RPPPPRVASV LGTMEMGRRM DAPASAAAVR AFLERGHTEL DTAFMYSDGQ SETILGGLGL GLGGGDCRVK IATKANPWDG KSLKPDSVRS QLETSLKRLQ CPQVDLFYLH APDHGTPVEE TLHACQRLHQ EGKFVELGLS NYASWEVAEI CTLCKSNGWI LPTVYQGMYN ATTRQVETEL FPCLRHFGLR FYAYNPLAGG LLTGKYKYED KDGKQPVGRF FGNSWAETYR NRFWKEHHFE AIALVEKALQ AAYGASAPSV TSAALRWMYH HSQLQGAHGD AVILGMSSLE QLEQNLAATE EGPLEPAVVD AFNQAWHLVA HECPNYFR | |
Greater than 90.0% as determined by SDS-PAGE. |