missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Aldose 1-epimerase Protein
A DNA sequence encoding the Homo sapiens (Human) Aldose 1-epimerase, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7961-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_620156.1 | |
Aldose 1-epimerase Protein | |
Research Use Only | |
GALM | |
Recombinant Protein | |
E. coli | |
ASVTRAVFGE LPSGGGTVEK FQLQSDLLRV DIISWGCTIT ALEVKDRQGR ASDVVLGFAE LEGYLQKQPY FGAVIGRVAN RIAKGTFKVD GKEYHLAINK EPNSLHGGVR GFDKVLWTPR VLSNGVQFSR ISPDGEEGYP GELKVWVTYT LDGGELIVNY RAQASQATPV NLTNHSYFNL AGQASPNIND HEVTIEADTY LPVDETLIPT GEVAPVQGTA FDLRKPVELG KHLQDFHLNG FDHNFCLKGS KEKHFCARVH HAASGRVLEV YTTQPGVQFY TGNFLDGTLK GKNGAVYPKH SGFCLETQNW PDAVNQPRFP PVLLRPGEEY DHTTWFKFSV A | |
Greater than 90% as determined by SDS-PAGE. |
130589 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |