missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Annexin A9 Protein
A DNA sequence encoding the Homo sapiens (Human) Annexin A9, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5655-ec-500ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_003559.2 | |
Annexin A9 Protein | |
Research Use Only | |
ANXA9 | |
Recombinant Protein | |
E. coli | |
MAPSLTQEIL SHLGLASKTA AWGTLGTLRT FLNFSVDKDA QRLLRAITGQ GVDRSAIVDV LTNRSREQRQ LISRNFQERT QQDLMKSLQA ALSGNLERIV MALLQPTAQF DAQELRTALK ASDSAVDVAI EILATRTPPQ LQECLAVYKH NFQVEAVDDI TSETSGILQD LLLALAKGGR DSYSGIIDYN LAEQDVQALQ RAEGPSREET WVPVFTQRNP EHLIRVFDQY QRSTGQELEE AVQNRFHGDA QVALLGLASV IKNTPLYFAD KLHQALQETE PNYQVLIRIL ISRCETDLLS IRAEFRKKFG KSLYSSLQDA VKGDCQSALL ALCRAEDM | |
Greater than 90% as determined by SDS-PAGE. |
8416 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |