missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human APO D Protein
A cDNA sequence encoding the APO D was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11042-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
347 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MVMLLLLLSA LAGLFGAAEG QAFHLGKCPN PPVQENFDVN KYLGRWYEIE KIPTTFENGR CIQANYSLME NGKIKVLNQE LRADGTVNQI EGEATPVNLT EPAKLEVKFS WFMPSAPYWI LATDYENYAL VYSCTCIIQL FHVDFAWILA RNPNLPPETV DSLKNILTSN NIDVKKMTVT DQVNCPKLS |
APO D Protein | |
Research Use Only | |
APOD | |
Human | |
Untagged | |
APO-D protein at 100microg/ml in 50mM Tris-Acetate, pH 7.5, 1mM EDTA and 20% Glycerol. |