missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human AS3MT Protein
A cDNA sequence encoding the AS3MT was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11080-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
AS3MT Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMAALRD AEIQKDVQTY YGQVLKRSAD LQTNGCVTTA RPVPKHIREA LQNVHEEVAL RYYGCGLVIP EHLENCWILD LGSGSGRDCY VLSQLVGEKG HVTGIDMTKG QVEVAEKYLD YHMEKYGFQA SNVTFIHGYI EKLGEAGIKN ESHDIVVSNCVINLVPDKQQ VLQEAYRVLK HGGELYFSDV YTSLELPEEI RTHKVLWGEC LGGALYWKEL AVLAQKIGFC PPRLVTANLI TIQNKELERV IGDCRFVSAT FRLFKHSKTG PTKRCQVIYN GGITGHEKEL MFDANFTFKE GEIVEVDEET AAILKNSRFA QDFLIRPIGE KLPTSGGCSA LELKDIITDP FKLAEESDSM KSRCVPDAAG GCCGTKKSC | |
Greater than 90.0% as determined by SDS-PAGE. |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
AS3MT protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.15M NaCl. |