missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human B7-H4 / B7S1 / B7x Protein
A DNA sequence encoding the Homo sapiens (Human) B7-H4 / B7S1 / B7x, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8751-ec-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001240778.1 | |
B7-H4 / B7S1 / B7x Protein | |
Research Use Only | |
VTCN1 | |
Recombinant Protein | |
E. coli | |
IIGFGISGRH SITVTTVASA GNIGEDGILS CTFEPDIKLS DIVIQWLKEG VLGLVHEFKE GKDELSEQDE MFRGRTAVFA DQVIVGNASL RLKNVQLTDA GTYKCYIITS KGKGNANLEY KTGAFSMPEV NVDYNASSET LRCEAPRWFP QPTVVWASQV DQGANFSEVS NTSFELNSEN VTMKVVSVLY NVTINNTYSC MIENDIAKAT GDIKVTESEI KRRSHLQLLN SKA | |
Greater than 90% as determined by SDS-PAGE. |
79679 | |
1 mg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |