missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BCL10 Protein
A DNA sequence encoding the Homo sapiens (Human) BCL10, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5709-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_003912.1 | |
BCL10 Protein | |
Research Use Only | |
BCL10 | |
Recombinant Protein | |
E. coli | |
MEPTAPSLTE EDLTEVKKDA LENLRVYLCE KIIAERHFDH LRAKKILSRE DTEEISCRTS SRKRAGKLLD YLQENPKGLD TLVESIRREK TQNFLIQKIT DEVLKLRNIK LEHLKGLKCS SCEPFPDGAT NNLSRSNSDE SNFSEKLRAS TVMYHPEGES STTPFFSTNS SLNLPVLEVG RTENTIFSST TLPRPGDPGA PPLPPDLQLE EEGTCANSSE MFLPLRSRTV SRQ | |
Greater than 90% as determined by SDS-PAGE. |
8915 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |