missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Beta Defensin 103AB Protein
A DNA sequence encoding the Human Beta Defensin 103AB, was expressed in the hosts and tags indicated.
Marke: enQuireBio™ QP10431-ec-5ug
Weitere Details : Peso : 0.01000kg
Spezifikation
NP_001075020.1 | |
Beta Defensin 103AB Protein | |
Research Use Only | |
DEFB103AB | |
The ED50 as determined by anti-microbial activity against E.coli. is less than 30 ug/ml, corresponding to a specific activity of > 33.3 IU/mg. | |
Human | |
Untagged | |
0.2 um filtered 20 mM PBS, pH 7.4, 130 mM NaCl, lyophilized |
414325 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GIINTLQKYY CRVRGGRCAV LSCLPKEEQI GKCSTRGRKC CRRKK | |
Greater than 98% as determined by SDS-PAGE and HPLC. |