missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BIRC7 Protein
A cDNA sequence encoding the BIRC7 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11164-HIS-EC-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
BIRC7 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGPKDSAKCL HRGPQPSHWA AGDGPTQERC GPRSLGSPVL GLDTCRAWDH VDGQILGQLR PLTEEEEEEG AGATLSRGPA FPGMGSEELRLASFYDWPLT AEVPPELLAA AGFFHTGHQD KVRCFFCYGG LQSWKRGDDP WTEHAKWFPS CQFLLRSKGR DFVHSVQETH SQLLGSWDPW EEPEDAAPVA PSVPASGYPE LPTPRREVQS ESAQEPGGVS PAEAQRAWWV LEPPGARDVE AQLRRLQEER TCKVCLDRAV SIVFVPCGHL VCAECAPGLQ LCPICRAPVR SRVRTFLS | |
Greater than 90.0% as determined by(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The BIRC7 solution contains 20mM Tris pH 8.0, 1mM DTT, 0.1M NaCl, 2mM EDTA and 20% glycerol. |