missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human BMP 5 Protein
A cDNA sequence encoding the BMP 5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11171-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
653 | |
5 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MAANKRKNQN RNKSSSHQDS SRMSSVGDYN TSEQKQACKK HELYVSFRDL GWQDWIIAPE GYAAFYCDGE CSFPLNAHMN ATNHAIVQTL VHLMFPDHVP KPCCAPTKLN AISVLYFDDS SNVILKKYRN MVVRSCGCH | |
Greater than 95.0% as determined by SDS-PAGE. |
BMP 5 Protein | |
Research Use Only | |
BMP5 | |
Human | |
Untagged | |
The BMP-5 solution contains 10mM Sodium Citrate buffer (pH3.5) and 10% Glycerol. |