missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-Myc-binding Protein
A DNA sequence encoding the Homo sapiens (Human) C-Myc-binding protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8016-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_036465.2 | |
C-Myc-binding Protein | |
Research Use Only | |
MYCBP | |
Recombinant Protein | |
E. coli | |
AHYKAADSKR EQFRRYLEKS GVLDTLTKVL VALYEEPEKP NSALDFLKHH LGAATPENPE IELLRLELAE MKEKYEAIVE ENKKLKAKLA QYEPPQEEKR AE | |
Greater than 90% as determined by SDS-PAGE. |
26292 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |