missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C-type lectin domain family 18 member A Protein
A DNA sequence encoding the Homo sapiens (Human) C-type lectin domain family 18 member A, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5837-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001129686.1 | |
C-type lectin domain family 18 member A Protein | |
Research Use Only | |
CLEC18A | |
Recombinant Protein | |
E. coli | |
EVWPPQLQEQ APMAGALNRK ESFLLLSLHN RLRSWVQPPA ADMRRLDWSD SLAQLAQARA ALCGTPTPSL ASGLWRTLQV GWNMQLLPAG LVSFVEVVSL WFAEGQRYSH AAGECARNAT CTHYTQLVWA TSSQLGCGRH LCSAGQAAIE AFVCAYSPRG NWEVNGKTIV PYKKGAWCSL CTASVSGCFK AWDHAGGLCE VPRNPCRMSC QNHGRLNIST CHCHCPPGYT GRYCQVRCSL QCVHGRFREE ECSCVCDIGY GGAQCATKVH FPFHTCDLRI DGDCFMVSSE ADTYYRARMK CQRKGGVLAQ IKSQKVQDIL AFYLGRLETT NEVIDSDFET RNFWIGLTYK TAKDSFRWAT GEHQAFTSFA FGQPDNHGFG NCVELQASAA FNWNDQRCKT RNRYICQFAQ EHISRWGPGS | |
Greater than 90% as determined by SDS-PAGE. |
348174 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |