missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human C9 Protein
A cDNA sequence encoding the C9 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11243-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
C9 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSQYTTSYD PELTESSGSA SHIDCRMSPW SEWSQCDPCL RQMFRSRSIE VFGQFNGKRC TDAVGDRRQC VPTEPCEDAE DDCGNDFQCS TGRCIKMRLR CNGDNDCGDF SDEDDCESEP RPPCRDRVVE ESELARTAGY GINILGMDPL STPFDNEFYN GLCNRDRDGN TLTYYRRPWN VASLIYETKG EKNFRTEHYE EQIEAFKSII QEKTSNFNAA ISLKFTPTET NKAEQCCEET ASSISLHGKG SFRFSYSKNE TYQLFLSYSS KKEKMFLHVK GEIHLGRFVM RNRDVVLTTT FVDDIKALPT TYEKGEYFAF LETYGTHYSS SGSLGGLYEL IYVLDKASMK RKGVELKDIK RCLGYHLDVS LAFSEISVGA EFNKDDCVKR GEGRAVNITS ENLIDDVVSL IRGGTRKYAF ELKEKLLRGT VIDVTDFVNW ASSINDAPVL ISQKLSPIYN LVPVKMKNAH LKKQNLERAI EDYINEFSVR KCHTCQNGGT VILMDGKCLC ACPFKFEGIA CEISKQKISE GLPALEFPNE K | |
Greater than 80% as determined by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
C9 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 10% glycerol and 0.4M Urea. |