missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CALB1 Protein
A cDNA sequence encoding the CALB1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11258-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CALB1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MAESHLQSSL ITASQFFEIW LHFDADGSGY LEGKELQNLI QELQQARKKA GLELSPEMKT FVDQYGQRDD GKIGIVELAH VLPTEENFLL LFRCQQLKSC EEFMKTWRKY DTDHSGFIET EELKNFLKDL LEKANKTVDD TKLAEYTDLM LKLFDSNNDG KLELTEMARL LPVQENFLLK FQGIKMCGKE FNKAFELYDQ DGNGYIDENE LDALLKDLCE KNKQDLDINN ITTYKKNIMA LSDGGKLYRT DLALILCAGD N | |
Greater than 95.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
Untagged | |
The CALB1 protein solution contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol and 2mM EDTA. |