missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCL11 / Eotaxin Protein
A DNA sequence encoding the Human CCL11 / Eotaxin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10232-ec-5ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002977.1 | |
CCL11 / Eotaxin Protein | |
Research Use Only | |
CCL11 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood eosinophils is in a concentration range of 0.1-10.0 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
6356 | |
5 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
GPASVPTTCC FNLANRKIPL QRLESYRRIT SGKCPQKAVI FKTKLAKDIC ADPKKKWVQD SMKYLDQKSP TPKP | |
Greater than 97% as determined by SDS-PAGE and HPLC. |