missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CCNA2 Protein
A cDNA sequence encoding the CCNA2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11294-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
890 | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLGNSA PGPATREAGS ALLALQQTAL QEDQENINPE KAAPVQQPRT RAALAVLKSG NPRGLAQQQR PKTRRVAPLK DLPVNDEHVT VPPWKANSKQ PAFTIHVDEA EKEAQKKPAE SQKIEREDAL AFNSAISLPG PRKPLVPLDY PMDGSFESPHTMDMSIVLED EKPVSVNEVP DYHEDIHTYL REMEVKCKPK VGYMKKQPDI TNSMRAILVD WLVEVGEEYK LQNETLHLAV NYIDRFLSSM SVLRGKLQLV GTAAMLLASK FEEIYPPEVA EFVYITDDTY TKKQVLRMEH LVLKVLTFDL AAPTVNQFLT QYFLHQQPAN CKVESLAMFLGELSLIDADP YLKYLPSVIA GAAFHLALYT VTGQSWPESL IRKTGYTLES LKPCLMDLHQ TYLKAPQHAQ QSIREKYKNS KYHGVSLLNP PETLNL | |
Greater than 90% as determined by SDS-PAGE. |
CCNA2 Protein | |
Research Use Only | |
CCNA2 | |
Human | |
His | |
CCNA2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.15M NaCl and 1mM DTT. |