missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD244 Protein
A cDNA sequence encoding the CD244 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11316-HIS-EC-1mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
51744 | |
Greater than 85% as determined by SDS-PAGE. | |
Research Use Only | |
CD244 | |
Human | |
His | |
CD244 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer, pH 8.0 and 0.4M Urea. |
CD244 Protein | |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHGKGCQG SADHVVSISG VPLQLQPNSI QTKVDSIAWK KLLPSQNGFH HILKWENGSL PSNTSNDRFS FIVKNLSLLI KAAQQQDSGL YCLEVTSISG KVQTATFQVF VFDKVEKPRL QGQGKILDRG RCQVALSCLV SRDGNVSYAW YRGSKLIQTA GNLTYLDEEV DINGTHTYTC NVSNPVSWES HTLNLTQDCQ NAHQEFRFWP |