missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD274 Protein
A cDNA sequence encoding the CD274 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11319-HIS-EC-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
29126 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMFTVTV PKDLYVVEYG SNMTIECKFP VEKQLDLAAL IVYWEMEDKN IIQFVHGEED LKVQHSSYRQ RARLLKDQLS LGNAALQITD VKLQDAGVYR CMISYGGADY KRITVKVNAP YNKINQRILV VDPVTSEHEL TCQAEGYPKA EVIWTSSDHQ VLSGKTTTTN SKREEKLFNV TSTLRINTTT NEIFYCTFRR LDPEENHTAE LVIPELPLAH PPNER | |
Greater than 90% as determined by SDS-PAGE. |
CD274 Protein | |
Research Use Only | |
CD274 | |
Human | |
His | |
The CD274 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 1mM DTT and 10% glycerol. |