missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD48 Protein
A cDNA sequence encoding the CD48 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11337-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
962 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
Baculovirus | |
ADPQGHLVHM TVVSGSNVTL NISESLPENY KQLTWFYTFD QKIVEWDSRK SKYFESKFKG RVRLDPQSGA LYISKVQKED NSTYIMRVLK KTGNEQEWKI KLQVLDPVPK PVIKIEKIED MDDNCYLKLS CVIPGESVNY TWYGDKRPFP KELQNSVLET TLMPHNYSRC YTCQVSNSVSSKNGTVCLSP PCTLARSHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
CD48 Protein | |
Research Use Only | |
CD48 | |
Human | |
His | |
NECTIN2 protein solution (1.0 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |