missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD70 / CD27L / TNFSF7 Protein
A DNA sequence encoding the Homo sapiens (Human) CD70 / CD27L / TNFSF7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9053-ye-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001243.1 | |
CD70 / CD27L / TNFSF7 Protein | |
Research Use Only | |
CD70 | |
Recombinant Protein | |
Yeast | |
QRFAQAQQQL PLESLGWDVA ELQLNHTGPQ QDPRLYWQGG PALGRSFLHG PELDKGQLRI HRDGIYMVHI QVTLAICSST TASRHHPTTL AVGICSPASR SISLLRLSFH QGCTIASQRL TPLARGDTLC TNLTGTLLPS RNTDETFFGV QWVRP | |
Greater than 90% as determined by SDS-PAGE. |
970 | |
10 μg | |
Not determined. | |
Extracellular Domain | |
Human | |
His | |
Tris-based buffer, 50% glycerol |