missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CD74 Protein
A cDNA sequence encoding the CD74 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11346-HIS-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
972 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
CD74 | |
Human | |
His | |
CD74 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 30% glycerol and 1mM DTT. |
CD74 Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. Coli | |
MGSSHHHHHH SSGLVPRGSH MGSQQQGRLD KLTVTSQNLQ LENLRMKLPK PPKPVSKMRM ATPLLMQALP MGALPQGPMQ NATKYGNMTE DHVMHLLQNA DPLKVYPPLK GSFPENLRHL KNTMETIDWK VFESWMHHWL LFEMSRHSLE QKPTDAPPKE SLELEDPSSG LGVTKQDLGP VPM |