missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CEACAM3 Protein
A cDNA sequence encoding the CEACAM3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11382-HIS-EC-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CEACAM3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSKLTIESM PLSVAEGKEV LLLVHNLPQH LFGYSWYKGE RVDGNSLIVG YVIGTQQATP GAAYSGRETI YTNASLLIQN VTQNDIGFYT LQVIKSDLVN EEATGQFHVY QENAPGLPVG AVAG | |
Greater than 85.0% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CEACAM3 protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4) and 20% glycerol. |