missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cell cycle checkpoint protein RAD1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cell cycle checkpoint protein RAD1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6577-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002844.1 | |
Cell cycle checkpoint protein RAD1 Protein | |
Research Use Only | |
RAD1 | |
Recombinant Protein | |
E. coli | |
MPLLTQQIQD EDDQYSLVAS LDNVRNLSTI LKAIHFREHA TCFATKNGIK VTVENAKCVQ ANAFIQAGIF QEFKVQEESV TFRINLTVLL DCLSIFGSSP MPGTLTALRM CYQGYGYPLM LFLEEGGVVT VCKINTQEPE ETLDFDFCST NVINKIILQS EGLREAFSEL DMTSEVLQIT MSPDKPYFRL STFGNAGSSH LDYPKDSDLM EAFHCNQTQV NRYKISLLKP STKALVLSCK VSIRTDNRGF LSLQYMIRNE DGQICFVEYY CCPDEEVPES ES | |
Greater than 80% as determined by SDS-PAGE. |
5810 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |