missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Centromere protein 1
A DNA sequence encoding the Homo sapiens (Human) Centromere protein H, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5818-ec-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_075060.1 | |
Centromere protein 1 | |
Research Use Only | |
CENPH | |
Recombinant Protein | |
E. coli | |
MEEQPQMQDA DEPADSGGEG RAGGPPQVAG AQAACSEDRM TLLLRLRAQT KQQLLEYKSM VDASEEKTPE QIMQEKQIEA KIEDLENEIE EVKVAFEIKK LALDRMRLST ALKKNLEKIS RQSSVLMDNM KHLLELNKLI MKSQQESWDL EEKLLDIRKK RLQLKQASES KLLEIQTEKN KQKIDLDSME NSERIKIIRQ NLQMEIKITT VIQHVFQNLI LGSKVNWAED PALKEIVLQL EKNVDMM | |
Greater than 80% as determined by SDS-PAGE. |
64946 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |