missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CGB3 Protein
A cDNA sequence encoding the CGB3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11399-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CGB3 Protein | |
Research Use Only | |
Recombinant Protein | |
Baculovirus | |
ADPSKEPLRP RCRPINATLA VEKEGCPVCI TVNTTICAGY CPTMTRVLQG VLPALPQVVC NYRDVRFESI RLPGCPRGVN PVVSYAVALS CQCALCRRST TDCGGPKDHP LTCDDPRFQD SSSSKAPPPS LPSPSRLPGP SDTPILPQHH HHHH | |
Greater than 90.0% as determined by SDS-PAGE. |
50 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CGB3 protein solution (0.25 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |