Learn More
enQuireBio™ Recombinant Human Coagulation Factor VII / FVII / F7 Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor VII / FVII / F7, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6012-ec-100ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000122.1 | |
Coagulation Factor VII / FVII / F7 Protein | |
Research Use Only | |
F7 | |
Recombinant Protein | |
E. coli | |
ANAFLEELRP GSLERECKEE QCSFEEAREI FKDAERTKLF WISYSDGDQC ASSPCQNGGS CKDQLQSYIC FCLPAFEGRN CETHKDDQLI CVNENGGCEQ YCSDHTGTKR SCRCHEGYSL LADGVSCTPT VEYPCGKIPI LEKRNASKPQ GR | |
Greater than 90% as determined by SDS-PAGE. |
2155 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |