missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Coagulation factor XIII A chain Protein

A DNA sequence encoding the Homo sapiens (Human) Coagulation factor XIII A chain, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9132-ye-1mg

Dettagli aggiuntivi : Peso : 0.01000kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16067691

Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.


Descrizione

Descrizione

Specifica

Specifica

NP_000120.2
Coagulation factor XIII A chain Protein
Research Use Only
F13A1
Recombinant Protein
Yeast
GVNLQEFLNV TSVHLFKERW DTNKVDHHTD KYENNKLIVR RGQSFYVQID FSRPYDPRRD LFRVEYVIGR YPQENKGTYI PVPIVSELQS GKWGAKIVMR EDRSVRLSIQ SSPKCIVGKF RMYVAVWTPY GVLRTSRNPE TDTYILFNPW CEDDAVYLDN EKEREEYVLN DIGVIFYGEV NDIKTRSWSY GQFEDGILDT CLYVMDRAQM DLSGRGNPIK VSRVGSAMVN AKDDEGVLVG SWDNIYAYGV PPSAWTGSVD ILLEYRSSEN PVRYGQCWVF AGVFNTFLRC LGIPARIVTN YFSAHDNDAN LQMDIFLEED GNVNSKLTKD SVWNYHCWNE AWMTRPDLPV GFGGWQAVDS TPQENSDGMY RCGPASVQAI KHGHVCFQFD APFVFAEVNS DLIYITAKKD GTHVVENVDA THIGKLIVTK QIGGDGMMDI TDTYKFQEGQ EEERLALETA LMYGAKKPLN TEGVMKSRSN VDMDFEVENA VLGKDFKLSI TFRNNSHNRY TITAYLSANI TFYTGVPKAE FKKETFDVTL EPLSFKKEAV LIQAGEYMGQ LLEQASLHFF VTARINETRD VLAKQKSTVL TIPEIIIKVR GTQVVGSDMT VTVQFTNPLK ETLRNVWVHL DGPGVTRPMK KMFREIRPNS TVQWEEVCRP WVSGHRKLIA SMSSDSLRHV YGELDVQIQR RPSM
Greater than 90% as determined by SDS-PAGE.
2162
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali