missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coagulation factor XIII A chain Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation factor XIII A chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP9132-ye-50ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_000120.2 | |
Coagulation factor XIII A chain Protein | |
Research Use Only | |
F13A1 | |
Recombinant Protein | |
Yeast | |
GVNLQEFLNV TSVHLFKERW DTNKVDHHTD KYENNKLIVR RGQSFYVQID FSRPYDPRRD LFRVEYVIGR YPQENKGTYI PVPIVSELQS GKWGAKIVMR EDRSVRLSIQ SSPKCIVGKF RMYVAVWTPY GVLRTSRNPE TDTYILFNPW CEDDAVYLDN EKEREEYVLN DIGVIFYGEV NDIKTRSWSY GQFEDGILDT CLYVMDRAQM DLSGRGNPIK VSRVGSAMVN AKDDEGVLVG SWDNIYAYGV PPSAWTGSVD ILLEYRSSEN PVRYGQCWVF AGVFNTFLRC LGIPARIVTN YFSAHDNDAN LQMDIFLEED GNVNSKLTKD SVWNYHCWNE AWMTRPDLPV GFGGWQAVDS TPQENSDGMY RCGPASVQAI KHGHVCFQFD APFVFAEVNS DLIYITAKKD GTHVVENVDA THIGKLIVTK QIGGDGMMDI TDTYKFQEGQ EEERLALETA LMYGAKKPLN TEGVMKSRSN VDMDFEVENA VLGKDFKLSI TFRNNSHNRY TITAYLSANI TFYTGVPKAE FKKETFDVTL EPLSFKKEAV LIQAGEYMGQ LLEQASLHFF VTARINETRD VLAKQKSTVL TIPEIIIKVR GTQVVGSDMT VTVQFTNPLK ETLRNVWVHL DGPGVTRPMK KMFREIRPNS TVQWEEVCRP WVSGHRKLIA SMSSDSLRHV YGELDVQIQR RPSM | |
Greater than 90% as determined by SDS-PAGE. |
2162 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |