missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coiled-coil domain-containing protein 1
A DNA sequence encoding the Homo sapiens (Human) Coiled-coil domain-containing protein 3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8059-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001269587.1 | |
Coiled-coil domain-containing protein 1 | |
Research Use Only | |
CCDC3 | |
Recombinant Protein | |
E. coli | |
CQLPSEWRPL SEGCRAELAE TIVYARVLAL HPEAPGLYNH LPWQYHAGQG GLFYSAEVEM LCDQAWGSML EVPAGSRLNL TGLGYFSCHS HTVVQDYSYF FFLRMDENYN LLPHGVNFQD AIFPDTQENR RMFSSLFQFS NCSQGQQLAT FSSDWEIQED SRLMCSSVQK ALFEEEDHVK KLQQKVATLE KRNRQLRERV KKVKRSLRQA RKKGRHLELA NQKLSEKLAA GALPHINARG PVRPPYLRG | |
Greater than 90% as determined by SDS-PAGE. |
83643 | |
200 μg | |
Not determined. | |
Full Length of Mature Protein | |
Human | |
His | |
Tris-based buffer, 50% glycerol |