missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COPE Protein
A cDNA sequence encoding the COPE was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11484-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
11316 | |
Greater than 90.0% as determined by SDS-PAGE. | |
Research Use Only | |
COPE | |
Human | |
His | |
COPE protein solution (0.25 mg/ml) containing Phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT. |
COPE Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMAPPAPG PASGGSGEVD ELFDVKNAFY IGSYQQCINE AQRVKLSSPE RDVERDVFLY RAYLAQRKFG VVLDEIKPSS APELQAVRMF ADYLAHESRR DSIVAELDRE MSRSVDVTNT TFLLMAASIY LHDQNPDAAL RALHQGDSLE CTAMTVQILL KLDRLDLARK ELKRMQDLDE DATLTQLATA WVSLATGGEK LQDAYYIFQE MADKCSPTLL LLNGQAACHM AQGRWEAAEG LLQEALDKDS GYPETLVNLI VLSQHLGKPP EVTNRYLSQL KDAHRSHPFI KEYQAKENDF DRLVLQYAPS A |