missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CREB3L2 Protein
A DNA sequence encoding the Homo sapiens (Human) CREB3L2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7764-ec-200ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_001240704.1 | |
CREB3L2 Protein | |
Research Use Only | |
CREB3L2 | |
Recombinant Protein | |
E. coli | |
MEVLESGEQG VLQWDRKLSE LSEPGDGEAL MYHTHFSELL DEFSQNVLGQ LLNDPFLSEK SVSMEVEPSP TSPAPLIQAE HSYSLCEEPR AQSPFTHITS DSFNDDEVES EKWYLSTDFP STSIKTEPVT DEPPPGLVPS VTLTITAIST PLEKEEPPLE MNTGVDSSCQ TIIPKIKLEP HEVDQFLNFS PKEGLSALPV SLWVMDMVSG STEREYGERA GMSLYHRCCS WLYEIALFLK NKNFASK | |
Greater than 80% as determined by SDS-PAGE. |
64764 | |
200 μg | |
Not determined. | |
Full Length of BC063666 | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |