missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRYM Protein
A cDNA sequence encoding the CRYM was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11524-10ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
CRYM Protein | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
CRYM was filtered (0.4microm) and lyophilized from 0.5 mg/ml in 20mM Tris buffer and 50mM NaCl, pH 7.5. |
Greater than 95.0% as determined by SDS-PAGE. | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS RVPAFLSAAE VEEHLRSSSL LIPPLETALA NFSSGPEGGV MQPVRTVVPV TKHRGYLGVM PAYSAAEDAL TTKLVTFYED RGITSVVPSH QATVLLFEPS NGTLLAVMDG NVITAKRTAA VSAIATKFLK PPSSEVLCIL GAGVQAYSHY EIFTEQFSFK EVRIWNRTKE NAEKFADTVQ GEVRVCSSVQ EAVAGADVII TVTLATEPIL FGEWVKPGAH INAVGASRPD WRELDDELMK EAVLYVDSQE AALKESGDVL LSGAEIFAEL GEVIKGVKPA HCEKTTVFKS LGMAVEDTVA AKLIYDSWSS GK |