missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL3 / GRO gamma Protein
A DNA sequence encoding the Human CXCL3 / GRO gamma, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10207-ec-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002081.2 | |
CXCL3 / GRO gamma Protein | |
Research Use Only | |
CXCL3 | |
The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected human 293 cells is in a concentration range of 10-100 ng/ml. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
2921 | |
2 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN | |
Greater than 97% as determined by SDS-PAGE and HPLC. |