missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL5 / ENA-78 Protein
Recombinant Protein
Marca: enQuireBio™ QP1019-1 mg
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
NP_002985.1 | |
CXCL5 / ENA-78 Protein | |
Research Use Only | |
CXCL5 | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |
6374 | |
1 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AAVLRELRCV CLQTTQGVHP KMISNLQVFA IGPQCSKVEV VASLKNGKEI CLDPEAPFLK KVIQKILDGG NKEN | |
Greater than 95% as determined by SDS-PAGE. |