missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclophilin C Protein
A cDNA sequence encoding the Cyclophilin C was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11580-2ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifica
5480 | |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MKHHHHHHASFRKRGPSVTA KVFFDVRIGD KDVGRIVIGL FGKVVPKTVE NFVALATGEK GYGYKGSKFH RVIKDFMIQG GDITTGDGTG GVSIYGETFP DENFKLKHYG IGWVSMANAG PDTNGSQFFI TLTKPTWLDG KHVVFGKVID GMTVVHSIEL QATDGHDRPL TNCSIINSGK IDVKTPFVVE IADW | |
Greater than 95.0% as determined by SDS-PAGE. |
Cyclophilin C Protein | |
Research Use Only | |
PPIC | |
Human | |
His | |
Cyclophilin-C was filtered (0.4 microm) and lyophilized in 20mM Tris buffer and 50mM NaCl, pH 7.5. |