missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DCTN2 (1-403) Protein
A cDNA sequence encoding the DCTN2 (1-403) was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP14013-HIS-EC-20ug
Dettagli aggiuntivi : Peso : 0.01000kg
Specifications
DCTN2 (1-403) Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMADPKYA DLPGIARNEP DVYETSDLPE DDQAEFDAEL EELTSTSVEH IIVNPNAAYD KFKDKRVGTK GLDFSDRIGK TKRTGYESGE YEMLGEGLGV KETPQQKYQR LLHEVQELTT EVEKIKTTVK ESATEEKLTP VLLAKQLAAL KQQLVASHLE KLLGPDAAIN LTDPDGALAK RLLLQLEATK NSKGGSGGKT TGTPPDSSLV TYELHSRPEQ DKFSQAAKVA ELEKRLTELE TAVRCDQDAQ NPLSAGLQGA CLMETVELLQ AKVSALDLAV LDQVEARLQS VLGKVNEIAK HKASVEDADT QSKVHQLYET IQRWSPIAST LPELVQRLVT IKQLHEQAMQ FGQLLTHLDT TQQMIANSLK DNTTLLTQVQ TTMRENLATV EGNFASIDER MKKLGK | |
Greater than 90.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
DCTN2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl (pH 8.0), 10% glycerol, 0.15M NaCl and 1mM DTT. |